Sonoco™ ThermoSafe U-tek™ Gel Pack
Phase Change Material (PCM) gel pack for shipping temperature-sensitive materials.
Supplier:  Sonoco™ 429
 View more versions of this product
Catalog No. 03-528-6
Explore available promotions
Description
Description
  • Gel will phase change at -1 deg. or -23 deg.
  • Maintains critical temperatures of serums, test samples, live or frozen tissues, organs, and pharmaceuticals for extended periods.
  • Laminated, metallized film exterior provides radiant heat barrier and resists punctures.
  • Thermal solutions for temperature critical packages during transit
  • condition coolant in freezer below -1 deg. or -23 deg. before shipments.
  • For frozen or refrigerated shipments.
  • Alternative to dry ice.
  • Can be paired with our Mini Mailer(440).
  • Available in several sizes
Specifications
Specifications
36/Cs.
0.75 in.
-1°C
6 in.
Laminated, Metallized Film
1.2 kg
10.16 cm
U-tek Phase Change Material
Ideal for products requiring frozen temperatures or extreme cold that may not tolerate dry ice or Carbon dioxide
1.9 cm
15.2 cm
8 oz.
12 in.
6 x 4 x 0.75 in.