Sonoco™ ThermoSafe U-tek™ Gel Pack
Phase Change Material (PCM) gel pack for shipping temperature-sensitive materials.
Supplier:  Sonoco™ TS412
 View more versions of this product
Catalog No. 03-528B
Explore available promotions
Description
Description
  • Gel will phase change at -1 deg. or -23 deg.
  • Maintains critical temperatures of serums, test samples, live or frozen tissues, organs, and pharmaceuticals for extended periods.
  • Laminated, metallized film exterior provides radiant heat barrier and resists punctures.
  • Thermal solutions for temperature critical packages during transit
  • condition coolant in freezer below -1 deg. or -23 deg. before shipments.
  • For frozen or refrigerated shipments.
  • Alternative to dry ice.
  • Can be paired with our Mini Mailer(440).
  • Available in several sizes
Specifications
Specifications
24/Cs.
1 in.
-1°C
6.75 in.
Laminated, Metallized Film
0.68 kg
5.7 cm
U-tek Phase Change Material
Ideal for products requiring frozen temperatures or extreme cold that may not tolerate dry ice or Carbon dioxide
2.54 cm
17 cm
12 oz.
10.25 in.
6.75 x 4 x 1 in.