Eisco™ Thermostat Model
Thermostat Model for demonstration of operation of thermostat.
Supplier: Eisco™ PH0403
Description
- Vertically mounted bimetallic strip used to open or close electrical circuit with change in temperature
- Consists of binding posts for connection to low current circuits and adjustable contacts
Specifications
Thermostat Model | |
9 to Undergraduate | |
Demonstration of operation of thermostat | |
Vertically Mounted Bimetallic Strip |