Learn More
Thermo Scientific™ Thiacloprid
Chemical Identifiers
CAS | 111988-49-9 |
---|---|
Molecular Formula | C10H9ClN4S |
Molecular Weight (g/mol) | 252.72 |
InChI Key | HOKKPVIRMVDYPB-UHFFFAOYSA-N |
IUPAC Name | ({3-[(6-chloropyridin-3-yl)methyl]-1,3-thiazolidin-2-ylidene}amino)formonitrile |
SMILES | ClC1=CC=C(CN2CCSC2=NC#N)C=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC464390010
|
Thermo Scientific™
464390010 |
1 g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC464392500
|
Thermo Scientific™
464392500 |
250 mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
111988-49-9 | |
252.72 | |
({3-[(6-chloropyridin-3-yl)methyl]-1,3-thiazolidin-2-ylidene}amino)formonitrile |
C10H9ClN4S | |
HOKKPVIRMVDYPB-UHFFFAOYSA-N | |
ClC1=CC=C(CN2CCSC2=NC#N)C=N1 |
Specifications
111988-49-9 | |
1 g | |
HOKKPVIRMVDYPB-UHFFFAOYSA-N | |
({3-[(6-chloropyridin-3-yl)methyl]-1,3-thiazolidin-2-ylidene}amino)formonitrile | |
252.72 |
C10H9ClN4S | |
Solubility aq. soln.: .. Other solubilities: insoluble in chloroform & methanol | |
ClC1=CC=C(CN2CCSC2=NC#N)C=N1 | |
252.72 | |
Thiacloprid |
Safety and Handling
GHS H Statement Toxic if swallowed.
Harmful if inhaled.
May cause drowsiness or dizziness.
Suspected of causing cancer.
May damage fertility.
May damage the unborn child.
Very toxic to aquatic life with long last
GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Danger
RUO â Research Use Only