missing translation for 'onlineSavingsMsg'
Learn More

Thiamine Hydrochloride Hydrate 98.0+%, TCI America™

$76.43 - $499.57

Chemical Identifiers

CAS 67-03-8
Molecular Formula C12H18Cl2N4OS
Molecular Weight (g/mol) 337.263
MDL Number MFCD00012780
InChI Key DPJRMOMPQZCRJU-UHFFFAOYSA-M
Synonym thiamine hydrochloride, vitamin b1 hydrochloride, thiamine hcl, aneurine hydrochloride, berin, thiamine chloride hydrochloride, trophite, thiamin chloride, thiamine dichloride, thiaminium chloride
PubChem CID 6202
ChEBI CHEBI:49105
IUPAC Name 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride
SMILES CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T018125G
View Documents
TCI America
T018125G
25 g N/A
Only null left
 
T0181100G
View Documents
TCI America
T0181100G
100 g N/A
Only null left
 
T0181500G
View Documents
TCI America
T0181500G
500 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

67-03-8
337.263
DPJRMOMPQZCRJU-UHFFFAOYSA-M
6202
2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride
C12H18Cl2N4OS
MFCD00012780
thiamine hydrochloride, vitamin b1 hydrochloride, thiamine hcl, aneurine hydrochloride, berin, thiamine chloride hydrochloride, trophite, thiamin chloride, thiamine dichloride, thiaminium chloride
CHEBI:49105
CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]

Specifications

67-03-8
C12H18Cl2N4OS
25 g
DPJRMOMPQZCRJU-UHFFFAOYSA-M
2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride
6202
337.26
Crystalline Powder
White
MFCD00012780
thiamine hydrochloride, vitamin b1 hydrochloride, thiamine hcl, aneurine hydrochloride, berin, thiamine chloride hydrochloride, trophite, thiamin chloride, thiamine dichloride, thiaminium chloride
CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]
337.263
CHEBI:49105
≥98.0% (HPLC,T)
Thiamine Hydrochloride Hydrate
Videos
Safety and Handling

Safety and Handling

RTECSNumber : XI7350000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'