Learn More
Thiamine Hydrochloride, Powder, MP Biomedicals
Thiamine (Vitamin B1) is one of the essential vitamins and is required for carbohydrate metabolism.
Supplier:  MP Biomedicals Inc 0210302891
 View more versions of this product
Catalog No. ICN-10302891
Only null left
Explore available promotions
Description
Description
Thiamine hydrochloride is used as a food-additive to add a brothy/meaty flavor to gravies or soups. It is also used as food supplement. Used in fluorometric determination of mercury.
Specifications
Specifications
Thiamine Hydrochloride
67-03-8
MFCD00012780
3851771
UIERGBJEBXXIGO-UHFFFAOYSA-N
3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-3H-1λ4,3-thiazol-1-ylium nitrate
10762
Powder
Food Supplement
C12H17ClN4OS·HCl
1 kg
Aneurine hydrochloride, Vitamin B1 hydrochloride, Vitamin B1 HCl
[O-][N+]([O-])=O.CC1=C(CCO)[S+]=CN1CC1=CN=C(C)N=C1N
337.263
337.3
Chemical Identifiers
67-03-8
337.263
UIERGBJEBXXIGO-UHFFFAOYSA-N
10762
C12H17ClN4OS·HCl
MFCD00012780
Aneurine hydrochloride, Vitamin B1 hydrochloride, Vitamin B1 HCl
[O-][N+]([O-])=O.CC1=C(CCO)[S+]=CN1CC1=CN=C(C)N=C1N
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
EINECSNumber : 200-641-8
storageNote1 : Store Desiccated
Recommended Storage : Store at room temperature.