missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thiazole-5-methanol, 95%, Thermo Scientific™
Chemical Identifiers
CAS | 38585-74-9 |
---|---|
Molecular Formula | C4H5NOS |
Molecular Weight (g/mol) | 115.15 |
InChI Key | WKBQQWDVVHGWDB-UHFFFAOYSA-N |
Synonym | 5-hydroxymethylthiazole, 5-hydroxymethyl thiazole, thiazol-5-ylmethanol, 5-thiazolemethanol, thiazole-5-methanol, thiazol-5-yl-methanol, thiazol-5-yl methanol, 5-thiazolylmethanol, 5-hydroxymethyl-1,3-thiazole |
PubChem CID | 2763216 |
IUPAC Name | 1,3-thiazol-5-ylmethanol |
SMILES | C1=C(SC=N1)CO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC455550050
|
Thermo Scientific Chemicals
455550050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
38585-74-9 | |
115.15 | |
5-hydroxymethylthiazole, 5-hydroxymethyl thiazole, thiazol-5-ylmethanol, 5-thiazolemethanol, thiazole-5-methanol, thiazol-5-yl-methanol, thiazol-5-yl methanol, 5-thiazolylmethanol, 5-hydroxymethyl-1,3-thiazole | |
1,3-thiazol-5-ylmethanol |
C4H5NOS | |
WKBQQWDVVHGWDB-UHFFFAOYSA-N | |
2763216 | |
C1=C(SC=N1)CO |
Specifications
38585-74-9 | |
94% min. (GC) | |
5g | |
5-hydroxymethylthiazole, 5-hydroxymethyl thiazole, thiazol-5-ylmethanol, 5-thiazolemethanol, thiazole-5-methanol, thiazol-5-yl-methanol, thiazol-5-yl methanol, 5-thiazolylmethanol, 5-hydroxymethyl-1,3-thiazole | |
C1=C(SC=N1)CO | |
115.15 | |
115.15 | |
Thiazole-5-methanol |
1.3200g/mL | |
C4H5NOS | |
1.32 | |
WKBQQWDVVHGWDB-UHFFFAOYSA-N | |
1,3-thiazol-5-ylmethanol | |
2763216 | |
95% |
Safety and Handling
GHS H Statement Causes serious eye damage. Harmful to aquatic life with long lasting effects.
GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
RUO â Research Use Only