missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thieno[3,2-b]pyridin-7-ol, 96%, Thermo Scientific Chemicals

$96.96 - $328.71

Chemical Identifiers

CAS 107818-20-2
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.183
MDL Number MFCD01863636
InChI Key AACVULYSNJAKEQ-UHFFFAOYSA-N
Synonym thieno 3,2-b pyridin-7-ol, thieno 3,2-b pyridin-7 4h-one, 7-hydroxythieno 3,2-b pyridine, 4h-thieno 3,2-b pyridin-7-one, thiopheno 3,2-b pyridin-7-ol, thieno 3 2-b pyridin-7-ol, pubchem22356, acmc-2098xl, ksc173k8p
PubChem CID 5074765
IUPAC Name 4H-thieno[3,2-b]pyridin-7-one
SMILES C1=CNC2=C(C1=O)SC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5017203
View Documents
Thermo Scientific Chemicals
H5017203
1 g
Each for $96.96
 
AAH5017206
View Documents
Thermo Scientific Chemicals
H5017206
5 g
Each for $328.71
 
Description

Description

Thieno[3,2-b]pyridin-7-ol is used for pharmaceutical intermediates, as drugs, food, cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thieno[3,2-b]pyridin-7-ol is used for pharmaceutical intermediates, as drugs, food, cosmetics.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

107818-20-2
151.183
AACVULYSNJAKEQ-UHFFFAOYSA-N
5074765
C1=CNC2=C(C1=O)SC=C2
C7H5NOS
MFCD01863636
thieno 3,2-b pyridin-7-ol, thieno 3,2-b pyridin-7 4h-one, 7-hydroxythieno 3,2-b pyridine, 4h-thieno 3,2-b pyridin-7-one, thiopheno 3,2-b pyridin-7-ol, thieno 3 2-b pyridin-7-ol, pubchem22356, acmc-2098xl, ksc173k8p
4H-thieno[3,2-b]pyridin-7-one

Specifications

107818-20-2
MFCD01863636
thieno 3,2-b pyridin-7-ol, thieno 3,2-b pyridin-7 4h-one, 7-hydroxythieno 3,2-b pyridine, 4h-thieno 3,2-b pyridin-7-one, thiopheno 3,2-b pyridin-7-ol, thieno 3 2-b pyridin-7-ol, pubchem22356, acmc-2098xl, ksc173k8p
AACVULYSNJAKEQ-UHFFFAOYSA-N
4H-thieno[3,2-b]pyridin-7-one
5074765
96%
C7H5NOS
1 g
Soluble in water.
C1=CNC2=C(C1=O)SC=C2
151.183
151.19
Thieno[3,2-b]pyridin-7-ol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only