missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thiobenzoic Acid 93.0+%, TCI America™
Supplier: TCI America T019525G
Specifications
Thiobenzoic Acid | |
87°C | |
MFCD00004852 | |
thiobenzoic acid, benzenecarbothioic acid, benzoyl thiol, monothiobenzoic acid, benzoic acid, thio, acido mercaptobenzoico, unii-gbg5rlo56n, ccris 8913, acido mercaptobenzoico italian, gbg5rlo56n | |
C1=CC=C(C=C1)C(=O)S | |
138.184 | |
138.18 | |
Liquid |
98-91-9 | |
C7H6OS | |
25 g | |
UIJGNTRUPZPVNG-UHFFFAOYSA-N | |
benzenecarbothioic S-acid | |
7414 | |
≥93.0% (T) |
Chemical Identifiers
98-91-9 | |
138.184 | |
UIJGNTRUPZPVNG-UHFFFAOYSA-N | |
7414 | |
C1=CC=C(C=C1)C(=O)S |
C7H6OS | |
MFCD00004852 | |
thiobenzoic acid, benzenecarbothioic acid, benzoyl thiol, monothiobenzoic acid, benzoic acid, thio, acido mercaptobenzoico, unii-gbg5rlo56n, ccris 8913, acido mercaptobenzoico italian, gbg5rlo56n | |
benzenecarbothioic S-acid |
Safety and Handling
EINECSNumber : (3)-1396
RTECSNumber : DH6839000
TSCA : Yes
Recommended Storage : Refrigerator