missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thioisonicotinamide, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 138770250

 View more versions of this product

Catalog No. AC138770250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Thioisonicotinamide
Authentic
Glass bottle
MFCD00006437
thioisonicotinamide, 4-pyridinecarbothioamide, isothionicotinamide, 4-pyridylthiocarboxamide, 4-pyridylthioamide, thio-4-pyridinecarboxamide, isonicotinic acid thioamide, pyridine-4-thioamide, 4-pyridinecarboxamide, thio, isonicotinamide, thio
C1=CN=CC=C1C(=S)N
138.19
138.19
2196-13-6
96% min. (HPLC)
C6H6N2S
25g
KPIIGXWUNXGGCP-UHFFFAOYSA-N
pyridine-4-carbothioamide
2723788
97%

Chemical Identifiers

2196-13-6
138.19
KPIIGXWUNXGGCP-UHFFFAOYSA-N
2723788
C1=CN=CC=C1C(=S)N
C6H6N2S
MFCD00006437
thioisonicotinamide, 4-pyridinecarbothioamide, isothionicotinamide, 4-pyridylthiocarboxamide, 4-pyridylthioamide, thio-4-pyridinecarboxamide, isonicotinic acid thioamide, pyridine-4-thioamide, 4-pyridinecarboxamide, thio, isonicotinamide, thio
pyridine-4-carbothioamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning

EINECSNumber : 218-592-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only