missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thiolactic Acid 97.0+%, TCI America™

$41.48 - $211.32

Chemical Identifiers

CAS 79-42-5
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.14
MDL Number MFCD00004862
InChI Key PMNLUUOXGOOLSP-UHFFFAOYNA-N
Synonym 2-mercaptopropionic acid, thiolactic acid, 2-mercaptopropanoic acid, 2-thiolactic acid, 2-thiolpropionic acid, alpha-mercaptopropionic acid, alpha-mercaptopropanoic acid, propionic acid, 2-mercapto, propanoic acid, 2-mercapto, fema no. 3180
PubChem CID 62326
ChEBI CHEBI:47872
IUPAC Name 2-sulfanylpropanoic acid
SMILES CC(S)C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M006025G
View Documents
TCI America
M006025G
25 g
Each for $41.48
 
M0060100G
View Documents
TCI America
M0060100G
100 g
Each for $73.08
 
M0060500G
View Documents
TCI America
M0060500G
500 g
Each for $211.32
 
Description

Description

Specifications

Chemical Identifiers

79-42-5
106.14
PMNLUUOXGOOLSP-UHFFFAOYNA-N
62326
2-sulfanylpropanoic acid
C3H6O2S
MFCD00004862
2-mercaptopropionic acid, thiolactic acid, 2-mercaptopropanoic acid, 2-thiolactic acid, 2-thiolpropionic acid, alpha-mercaptopropionic acid, alpha-mercaptopropanoic acid, propionic acid, 2-mercapto, propanoic acid, 2-mercapto, fema no. 3180
CHEBI:47872
CC(S)C(O)=O

Specifications

79-42-5
115°C
MFCD00004862
2936
PMNLUUOXGOOLSP-UHFFFAOYNA-N
2-sulfanylpropanoic acid
62326
106.14
Liquid
Colorless
C3H6O2S
25 g
2-mercaptopropionic acid, thiolactic acid, 2-mercaptopropanoic acid, 2-thiolactic acid, 2-thiolpropionic acid, alpha-mercaptopropionic acid, alpha-mercaptopropanoic acid, propionic acid, 2-mercapto, propanoic acid, 2-mercapto, fema no. 3180
CC(S)C(O)=O
106.14
CHEBI:47872
≥97.0% (GC)
Thiolactic Acid
Videos
Safety and Handling

Safety and Handling

RTECSNumber : UF5250000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'