Learn More
Thionyl bromide, 97%
CAS: 507-16-4 | Br2OS | 207.87 g/mol
$114.74 - $305.40
Chemical Identifiers
| CAS | 507-16-4 |
|---|---|
| Molecular Formula | Br2OS |
| Molecular Weight (g/mol) | 207.87 |
| MDL Number | MFCD00011448 |
| InChI Key | HFRXJVQOXRXOPP-UHFFFAOYSA-N |
| Synonym | thionyl bromide, thionyl dibromide, unii-n6cu78b13t, thionylbromide, sulfur oxy dibromide, acmc-20ajin, pubchem2023, sobr2, thionyl bromide, purum at |
| PubChem CID | 68176 |
| IUPAC Name | sulfurooyl dibromide |
| SMILES | BrS(Br)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1564814
|
Thermo Scientific Chemicals
L1564814 |
25 g |
Each for $114.74
|
|
|||||
|
AAL1564822
|
Thermo Scientific Chemicals
L1564822 |
100 g |
Each for $305.40
|
|
|||||
Description
Thionyl bromide is used as a brominating agent in organic synthesis. It is used in the bromination of alfa-beta-unsaturated carbonyl compounds. It is also used to prepare alkyl bromides from alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThionyl bromide is used as a brominating agent in organic synthesis. It is used in the bromination of alfa-beta-unsaturated carbonyl compounds. It is also used to prepare alkyl bromides from alcohols.
Solubility
Miscible with benzene, toluene and ether.
Notes
Moisture and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water, hydrobromide, acetone and alcohol.It hydrolyzes readily to release hydrogen bromide.
Chemical Identifiers
| 507-16-4 | |
| 207.87 | |
| HFRXJVQOXRXOPP-UHFFFAOYSA-N | |
| 68176 | |
| BrS(Br)=O |
| Br2OS | |
| MFCD00011448 | |
| thionyl bromide, thionyl dibromide, unii-n6cu78b13t, thionylbromide, sulfur oxy dibromide, acmc-20ajin, pubchem2023, sobr2, thionyl bromide, purum at | |
| sulfurooyl dibromide |
Specifications
| 47°C to 49°C (20mmHg) | |
| 507-16-4 | |
| Br2OS | |
| UN3264 | |
| thionyl bromide, thionyl dibromide, unii-n6cu78b13t, thionylbromide, sulfur oxy dibromide, acmc-20ajin, pubchem2023, sobr2, thionyl bromide, purum at | |
| HFRXJVQOXRXOPP-UHFFFAOYSA-N | |
| sulfurooyl dibromide | |
| 68176 | |
| 97% | |
| 1.675 | |
| 2.683 |
| -52°C | |
| 25 g | |
| MFCD00011448 | |
| 14,9347 | |
| Miscible with benzene,toluene and ether. | |
| BrS(Br)=O | |
| 207.87 | |
| 207.88 | |
| Acrid | |
| Light and moisture sensitive | |
| Thionyl bromide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H312-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: I
EINECSNumber : 208-064-3
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only