missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thionyl chloride, 1M soln. in dichloromethane, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H31857AE

 View more versions of this product

Catalog No. AAH31857AE


Explore available promotions

Description

Description

Sulfuryl chloride and thionyl chloride have been found to be new chlorinating agents for alkyl sulfides. Preparation of aromatic polyesters by direct polycondensation with thionyl chloride in pyridine. Conversion of aromatic and. alpha.,. beta.-unsaturated aldehydes to dichlorides by thionyl chloride and dimethylformamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Thionyl chloride
Cl3P
MFCD00011449
1M soln. in dichloromethane
phosphorus trichloride, phosphorous trichloride, trichlorophosphine, phosphorus chloride, phosphorous chloride, phosphorus iii chloride, pcl3, phosphorus chloride pcl3, phosphine, trichloro, fosfortrichloride
FAIAAWCVCHQXDN-UHFFFAOYSA-N
trichlorophosphane
24387
118.97
Moisture sensitive; May form a precipitate
7719-09-7
SOCl2
UN2922
1.35
Reacts with water.
P(Cl)(Cl)Cl
137.32
CHEBI:30334
100 mL

Chemical Identifiers

7719-09-7
MFCD00011449
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H351
Causes severe skin burns and eye damage.
Suspected of causing cancer.

P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335-H350

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

EINECSNumber : 231-748-8

RTECSNumber : XM5150000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only