missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thiophene-2-carboxaldehyde, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1434436

 View more versions of this product

Catalog No. AAA1434436


Explore available promotions

Description

Description

Thiophene-2-carboxaldehyde is used in the preparation of beta-aryl-beta-amino acids and urea derivatives. It acts as an arylation reagent. Further, it is used in the synthesis of unsaturated ketones, which serve as antiviral and cytotoxic agents. In addition to this, it is used as an intermediate to manufacture pharmaceuticals and aromatic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene-2-carboxaldehyde is used in the preparation of beta-aryl-beta-amino acids and urea derivatives. It acts as an arylation reagent. Further, it is used in the synthesis of unsaturated ketones, which serve as antiviral and cytotoxic agents. In addition to this, it is used as an intermediate to manufacture pharmaceuticals and aromatic compounds.

Solubility
Miscible with chloroform, alcohol, ether, dimethylsulfoxide, ethyl acetate and most organic solvents. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
Specifications

Specifications

Thiophene-2-carboxaldehyde
1.22
78°C (172°F)
1.591
500 g
Air sensitive
Miscible with chloroform,alcohol,ether,dimethylsulfoxide,ethyl acetate and most organic solvents. Immiscible with water.
O=CC1=CC=CS1
112.15
CHEBI:87301
≥98%
98-03-3
196°C to 198°C
C5H4OS
MFCD00005429
105819
2-thiophenecarboxaldehyde, 2-thenaldehyde, thiophene-2-aldehyde, 2-formylthiophene, 2-thienylaldehyde, 2-thiophenealdehyde, thiophene-2-carboxaldehyde, 2-thienylcarboxaldehyde, 2-thienaldehyde, alpha-formylthiophene
CNUDBTRUORMMPA-UHFFFAOYSA-N
thiophene-2-carbaldehyde
7364
112.15

Chemical Identifiers

98-03-3
112.15
CNUDBTRUORMMPA-UHFFFAOYSA-N
7364
thiophene-2-carbaldehyde
C5H4OS
MFCD00005429
2-thiophenecarboxaldehyde, 2-thenaldehyde, thiophene-2-aldehyde, 2-formylthiophene, 2-thienylaldehyde, 2-thiophenealdehyde, thiophene-2-carboxaldehyde, 2-thienylcarboxaldehyde, 2-thienaldehyde, alpha-formylthiophene
CHEBI:87301
O=CC1=CC=CS1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H227-H319
Toxic if swallowed.
Combustible liquid.
Causes serious eye irritation.

P210-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P370+P378q-P501c

H227-H302-H315-H317-H319

EINECSNumber : 202-629-8

RTECSNumber : XM8135000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only