Learn More
Thiophene-2-carboxylic acid, 99%
CAS: 527-72-0 | C5H4O2S | 128.15 g/mol
$47.49 - $414.85
Chemical Identifiers
| CAS | 527-72-0 |
|---|---|
| Molecular Formula | C5H4O2S |
| Molecular Weight (g/mol) | 128.15 |
| MDL Number | MFCD00005437 |
| InChI Key | QERYCTSHXKAMIS-UHFFFAOYSA-N |
| Synonym | 2-thiophenecarboxylic acid, 2-thenoic acid, 2-carboxythiophene, 2-thiophenic acid, 2-thienylcarboxylic acid, alpha-thiophenecarboxylic acid, 2-tca, thiophene-2-carboxylate, tenoic acid, .alpha.-thiophenecarboxylic acid |
| PubChem CID | 10700 |
| ChEBI | CHEBI:71241 |
| IUPAC Name | thiophene-2-carboxylic acid |
| SMILES | OC(=O)C1=CC=CS1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1251414
|
Thermo Scientific Chemicals
A1251414 |
25 g |
Each for $47.49
|
|
|||||
|
AAA1251422
|
Thermo Scientific Chemicals
A1251422 |
100 g |
Each for $123.04
|
|
|||||
|
AAA1251436
|
Thermo Scientific Chemicals
A1251436 |
500 g |
Each for $414.85
|
|
|||||
Description
A hypoglycemic, anti-lipolytic agent
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 527-72-0 | |
| 128.15 | |
| QERYCTSHXKAMIS-UHFFFAOYSA-N | |
| 10700 | |
| thiophene-2-carboxylic acid |
| C5H4O2S | |
| MFCD00005437 | |
| 2-thiophenecarboxylic acid, 2-thenoic acid, 2-carboxythiophene, 2-thiophenic acid, 2-thienylcarboxylic acid, alpha-thiophenecarboxylic acid, 2-tca, thiophene-2-carboxylate, tenoic acid, .alpha.-thiophenecarboxylic acid | |
| CHEBI:71241 | |
| OC(=O)C1=CC=CS1 |
Specifications
| 527-72-0 | |
| 1.42 | |
| C5H4O2S | |
| 25 g | |
| 14,9354 | |
| QERYCTSHXKAMIS-UHFFFAOYSA-N | |
| thiophene-2-carboxylic acid | |
| 10700 | |
| 128.15 | |
| Thiophene-2-carboxylic acid |
| 126°C to 130°C | |
| 259°C to 261°C | |
| MFCD00005437 | |
| 110150 | |
| 2-thiophenecarboxylic acid, 2-thenoic acid, 2-carboxythiophene, 2-thiophenic acid, 2-thienylcarboxylic acid, alpha-thiophenecarboxylic acid, 2-tca, thiophene-2-carboxylate, tenoic acid, .alpha.-thiophenecarboxylic acid | |
| OC(=O)C1=CC=CS1 | |
| 128.15 | |
| CHEBI:71241 | |
| 99% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H319-H335
EINECSNumber : 208-423-4
RTECSNumber : XM8330200
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only