missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
S-Curve™ Three Compartment Glove Dispenser with Single Lid
Three compartment dispenser includes a single, sloping lid with a full-length hinge to maintain HEPA flow.
$544.08 - $776.75
Specifications
Product Type | Glove Dispenser |
---|---|
Color | Clear |
Holds | Gloves |
Mounting | Keyhole Slots |
No. of Compartments | 3 |
Catalog Number | Mfr. No. | Material | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity | |||||
19-160-197
|
S-Curve™
GD305 |
Acrylic |
Each for $544.08
|
|
|||||
19-162-425
|
S-Curve™
GD305PETG |
PETG |
Each for $776.75
|
|
|||||
Description
- Single, sloping lid with full-length hinge to maintain HEPA flow
- Can be mounted with heavy-duty wall brackets (included)
- Flame treated edges for a smooth finish
Specifications
Glove Dispenser | |
Gloves | |
3 | |
Heavy-duty wall mounting bracket |
Clear | |
Keyhole Slots | |
Top Loading | |
0.25 in. |