Thermo Scientific™ Three60™ Sampling System
Allows you to take multiple representative samples of your process liquids conveniently and in a closed system.
$1383.42 - $1387.33
Specifications
Content And Storage Store at room temperature.
Film Description ASI™ 77
Outer Packaging Placed into a thermoform tray and sealed
Port Detail 2 Ports (1 Luer Lock Insert + Cap, 1 Injection Site)
Material Ethyl Vinyl Acetate
View More Specs
Products 6
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
4MP0034
View Documents
Thermo Scientific™
4MP0034
50 mL
Case of 8 Each for $1,387.33
4MP0035
View Documents
Thermo Scientific™
4MP0035
100 mL
Case of 8 Each for $1,383.42
4MP0036
View Documents
Thermo Scientific™
4MP0036
250 mL
Case of 8 Each for $1,387.33
4MP0037
View Documents
Thermo Scientific™
4MP0037
500 mL
Case of 8 Each for $1,383.42
4MP0038
View Documents
Thermo Scientific™
4MP0038
1 L
Case of 8 Each for $1,383.42
4MP0039
View Documents
Thermo Scientific™
4MP0039
2 L
Case of 8 Each for $1,383.42
Description
Description
The Thermo Scientific™ Three60™ Sampling System allows you to take a representative sample of your product with minimal effort. The fluid paths of the four pre-irradiated BioProcess Containers (BPCs) and assemblies feature quick-turn valves and pinch-and-cut disconnections for fluid transfer and removal. The Three60 Sampling System is composed of Thermo Scientific™ ASI™ 77 film, a 5-layer, 7mL, polyethylene film and includes 2 ports (1 luer lock insert + cap, 1 injection site).
  • Compatible with a 3.8 cm (1.5-inch) sanitary connector
  • Requires no expensive hardware
  • Each Three60 package contains a valve and four assemblies with pinch-and-cut disconnections
  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
Specifications
Specifications
Store at room temperature.
Placed into a thermoform tray and sealed
Ethyl Vinyl Acetate
Room Temperature
Three60™
ASI™ 77
2 Ports (1 Luer Lock Insert + Cap, 1 Injection Site)
8/Cs.
Irradiated
Sampling System
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'