missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thymine, >99%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210306025
Specifications
Thymine | |
C5H6N2O2 | |
thymine, 5-methyluracil, thymin, 2,4-dihydroxy-5-methylpyrimidine, thymine anhydrate, 2,4 1h,3h-pyrimidinedione, 5-methyl, 5-methylpyrimidine-2,4 1h,3h-dione, 5-methyl-2,4 1h,3h-pyrimidinedione, thymin purine base, 5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione | |
CC1=CNC(=O)NC1=O | |
126.115 | |
CHEBI:17821 | |
>99% |
65-71-4 | |
25 g | |
RWQNBRDOKXIBIV-UHFFFAOYSA-N | |
5-methyl-1H-pyrimidine-2,4-dione | |
1135 | |
126.1 | |
Crystalline |
Chemical Identifiers
65-71-4 | |
126.115 | |
thymine, 5-methyluracil, thymin, 2,4-dihydroxy-5-methylpyrimidine, thymine anhydrate, 2,4 1h,3h-pyrimidinedione, 5-methyl, 5-methylpyrimidine-2,4 1h,3h-dione, 5-methyl-2,4 1h,3h-pyrimidinedione, thymin purine base, 5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione | |
CHEBI:17821 | |
CC1=CNC(=O)NC1=O |
C5H6N2O2 | |
RWQNBRDOKXIBIV-UHFFFAOYSA-N | |
1135 | |
5-methyl-1H-pyrimidine-2,4-dione |