Learn More
Thymol, 99%
CAS: 89-83-8 | C10H14O | 150.22 g/mol
$92.36 - $685.88
Chemical Identifiers
| CAS | 89-83-8 |
|---|---|
| Molecular Formula | C10H14O |
| Molecular Weight (g/mol) | 150.22 |
| MDL Number | MFCD00002309 |
| InChI Key | MGSRCZKZVOBKFT-UHFFFAOYSA-N |
| Synonym | thymol, 2-isopropyl-5-methylphenol, 5-methyl-2-isopropylphenol, thyme camphor, 3-p-cymenol, 6-isopropyl-m-cresol, thymic acid, isopropyl cresol, 5-methyl-2-1-methylethyl phenol, 6-isopropyl-3-methylphenol |
| PubChem CID | 6989 |
| ChEBI | CHEBI:27607 |
| IUPAC Name | 5-methyl-2-propan-2-ylphenol |
| SMILES | CC1=CC(=C(C=C1)C(C)C)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC150331000
|
Thermo Scientific Chemicals
150331000 |
100 g | Plastic bottle |
Each for $92.36
|
|
||||
|
AC150335000
|
Thermo Scientific Chemicals
150335000 |
500 g | Plastic bottle |
Each for $240.86
|
|
||||
|
AC150330025
|
Thermo Scientific Chemicals
150330025 |
2.5 kg | Plastic bottle |
Each for $685.88
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 89-83-8 | |
| 150.22 | |
| MGSRCZKZVOBKFT-UHFFFAOYSA-N | |
| 6989 | |
| 5-methyl-2-propan-2-ylphenol |
| C10H14O | |
| MFCD00002309 | |
| thymol, 2-isopropyl-5-methylphenol, 5-methyl-2-isopropylphenol, thyme camphor, 3-p-cymenol, 6-isopropyl-m-cresol, thymic acid, isopropyl cresol, 5-methyl-2-1-methylethyl phenol, 6-isopropyl-3-methylphenol | |
| CHEBI:27607 | |
| CC1=CC(=C(C=C1)C(C)C)O |
Specifications
| 89-83-8 | |
| White | |
| 232.0°C | |
| Authentic | |
| Plastic bottle | |
| MFCD00002309 | |
| 06, 532 | |
| 0.96 | |
| thymol, 2-isopropyl-5-methylphenol, 5-methyl-2-isopropylphenol, thyme camphor, 3-p-cymenol, 6-isopropyl-m-cresol, thymic acid, isopropyl cresol, 5-methyl-2-1-methylethyl phenol, 6-isopropyl-3-methylphenol | |
| MGSRCZKZVOBKFT-UHFFFAOYSA-N | |
| 5-methyl-2-propan-2-ylphenol | |
| 6989 | |
| CHEBI:27607 | |
| 99% | |
| Thymol, 99% |
| 48.0°C to 52.0°C | |
| 0.9600g/mL | |
| 102°C | |
| 98.5% min. (GC) | |
| C10H14O | |
| 100 g | |
| 0.05% max. | |
| 15, 9554 | |
| Solubility in water: 0.1% (20°C). Other solubilities: soluble in glacial acetic acid, soluble in alcohol, carbon disulfide, chloroform, and ether, slightly soluble in glycerol | |
| CC1=CC(=C(C=C1)C(C)C)O | |
| 150.22 | |
| 4.3 mm2/s (50°C) | |
| 150.22 | |
| Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement:
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 201-944-8
RTECSNumber : XP2275000
TSCA : TSCA
RUO – Research Use Only