missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thymol 99.0+%, TCI America™

Supplier:  TCI America M041025G

 View more versions of this product

Catalog No. M041025G


Explore available promotions

Description

Description

Specifications

Specifications

Thymol
51°C
232°C
MFCD00002309
1759
MGSRCZKZVOBKFT-UHFFFAOYSA-N
5-methyl-2-propan-2-ylphenol
6989
150.22
Crystalline Lumps
89-83-8
White
C10H14O
25 g
thymol, 2-isopropyl-5-methylphenol, 5-methyl-2-isopropylphenol, thyme camphor, 3-p-cymenol, 6-isopropyl-m-cresol, thymic acid, isopropyl cresol, 5-methyl-2-1-methylethyl phenol, 6-isopropyl-3-methylphenol
CC1=CC(=C(C=C1)C(C)C)O
150.221
CHEBI:27607
≥99.0% (GC)

Chemical Identifiers

89-83-8
150.221
MGSRCZKZVOBKFT-UHFFFAOYSA-N
6989
5-methyl-2-propan-2-ylphenol
C10H14O
MFCD00002309
thymol, 2-isopropyl-5-methylphenol, 5-methyl-2-isopropylphenol, thyme camphor, 3-p-cymenol, 6-isopropyl-m-cresol, thymic acid, isopropyl cresol, 5-methyl-2-1-methylethyl phenol, 6-isopropyl-3-methylphenol
CHEBI:27607
CC1=CC(=C(C=C1)C(C)C)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0521&(4)-0057

RTECSNumber : XP2275000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions