missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Thymol | |
51°C | |
232°C | |
MFCD00002309 | |
1759 | |
MGSRCZKZVOBKFT-UHFFFAOYSA-N | |
5-methyl-2-propan-2-ylphenol | |
6989 | |
150.22 | |
Crystalline Lumps |
89-83-8 | |
White | |
C10H14O | |
500 g | |
thymol, 2-isopropyl-5-methylphenol, 5-methyl-2-isopropylphenol, thyme camphor, 3-p-cymenol, 6-isopropyl-m-cresol, thymic acid, isopropyl cresol, 5-methyl-2-1-methylethyl phenol, 6-isopropyl-3-methylphenol | |
CC1=CC(=C(C=C1)C(C)C)O | |
150.221 | |
CHEBI:27607 | |
≥99.0% (GC) |
Chemical Identifiers
89-83-8 | |
150.221 | |
MGSRCZKZVOBKFT-UHFFFAOYSA-N | |
6989 | |
5-methyl-2-propan-2-ylphenol |
C10H14O | |
MFCD00002309 | |
thymol, 2-isopropyl-5-methylphenol, 5-methyl-2-isopropylphenol, thyme camphor, 3-p-cymenol, 6-isopropyl-m-cresol, thymic acid, isopropyl cresol, 5-methyl-2-1-methylethyl phenol, 6-isopropyl-3-methylphenol | |
CHEBI:27607 | |
CC1=CC(=C(C=C1)C(C)C)O |
Safety and Handling
EINECSNumber : (3)-0521&(4)-0057
RTECSNumber : XP2275000
TSCA : Yes