missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thymolphthalein, ACS Grade, LabChem™
Supplier: LabChem LC260607
Specifications
Thymolphthalein | |
100 | |
White | |
C28H30O4 | |
5 g | |
LDKDGDIWEUUXSH-UHFFFAOYSA-N | |
3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one | |
31316 | |
ACS |
125-20-2 | |
Carbon monoxide; Carbon dioxide | |
Amber Glass | |
C28H30O4 | |
thymolphthalein, thymophthalein, unii-yg5i28wsqp, yg5i28wsqp, phenolphthalein, 5',5-diisopropyl-2',2-dimethyl, 3,3-bis 4-hydroxy-5-isopropyl-2-methylphenyl-2-benzofuran-1 3h-one, 1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-2-methyl-5-1-methylethyl phenyl, 5',5-diisopropyl-2',2-dimethylphenolphthalein, 3,3-bis 4-hydroxy-2-methyl-5-propan-2-ylphenyl-2-benzofuran-1-one | |
CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O | |
430.544 | |
430.5 | |
Powder |
Chemical Identifiers
125-20-2 | |
430.544 | |
thymolphthalein, thymophthalein, unii-yg5i28wsqp, yg5i28wsqp, phenolphthalein, 5',5-diisopropyl-2',2-dimethyl, 3,3-bis 4-hydroxy-5-isopropyl-2-methylphenyl-2-benzofuran-1 3h-one, 1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-2-methyl-5-1-methylethyl phenyl, 5',5-diisopropyl-2',2-dimethylphenolphthalein, 3,3-bis 4-hydroxy-2-methyl-5-propan-2-ylphenyl-2-benzofuran-1-one | |
3,3-bis(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-2-benzofuran-1-one |
C28H30O4 | |
LDKDGDIWEUUXSH-UHFFFAOYSA-N | |
31316 | |
CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O |
Safety and Handling
GHS H Statement
Product is not hazardous.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage : Room Temperature