missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Thymopentin, 99+%

Reduces endocrine responses

$216.90 - $485.25

Chemical Identifiers

CAS 69558-55-0
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61431MB
View Documents
Thermo Scientific Chemicals
J61431MB
25mg
Each for $216.90
 
AAJ61431MC
View Documents
Thermo Scientific™
J61431MC
100mg
Each for $485.25
 
Description

Description

Reduces endocrine responses

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

69558-55-0

Specifications

≥99%
White
Powder
Thymopentin
69558-55-0
Store at -20°C
679.776
25mg
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only