Learn More
Tin(II) bromide, 99.2%, Thermo Scientific Chemicals
$45.56 - $154.59
Chemical Identifiers
CAS | 10031-24-0 |
---|---|
Molecular Formula | SnBr2 |
MDL Number | MFCD00011239 |
Synonym | tin ii bromide, tin dibromide, stannous bromide, tin bromide snbr2, snbr2, acmc-1bp22 |
Description
Stannous bromide acts a lewis acid. It is used to prepare alkyl tin compounds and in the spectrophotometric determination of iridium. It is involved in regioselective mono-etherfications of vicinal diols with diazo compounds. It acts as a catalyst to produce resinous lactone high polymers. It serves as a potent reducing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10031-24-0 | |
SnBr2 | |
MFCD00011239 | |
tin ii bromide, tin dibromide, stannous bromide, tin bromide snbr2, snbr2, acmc-1bp22 |
Specifications
623°C | |
10031-24-0 | |
5 g | |
SnBr2 | |
tin ii bromide, tin dibromide, stannous bromide, tin bromide snbr2, snbr2, acmc-1bp22 | |
278.51 | |
Tin(II) bromide |
215°C | |
Powder | |
99.4% | |
MFCD00011239 | |
Soluble in alcohol,ether,acetone,pyridine and dimethyl sulfoxide. | |
5.12 |
Safety and Handling
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 233-087-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only