missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) chloride, anhydrous, 99% min

CAS: 7772-99-8 | Cl2Sn | 189.61 g/mol

$188.89 - $2626.78

Chemical Identifiers

CAS 7772-99-8
Molecular Formula Cl2Sn
Molecular Weight (g/mol) 189.61
MDL Number MFCD00011241
InChI Key AXZWODMDQAVCJE-UHFFFAOYSA-L
Synonym Stannous chloride
PubChem CID 24479
ChEBI CHEBI:78067
SMILES [Cl-].[Cl-].[Sn++]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4196030
View Documents
Thermo Scientific Chemicals
04196030
250 g
Each for $188.89
Only null left
 
AA41960A1
View Documents
Thermo Scientific Chemicals
041960A1
1 kg
Each for $621.38
Only null left
 
AA41960A7
View Documents
Thermo Scientific Chemicals
041960A7
5 kg
Each for $2,626.78
Only null left
 
Description

Description

Tin(II) chloride is widely used as a reducing agent in organic synthesis, for example conversion of quinones into hydroquinones, nitroaromatics into aromatic amines, and in Stephen reduction. It acts as an antisludging agent and a stabilizer. It can be used to reduce certain metal salts into metals, tin plating and in analytical chemistry to test mercury ions. It is also used as a mordant in textile dyeing and as a catalyst in the production of polylactic acid. It is used as a food additive, wherein it serves as an anti-oxidant and a color retention agent. It can be used in the condensation of aryl aldehydes with cyclohexan-1,3-diones to synthesize xanthenes. It catalyzes direct conversion of aldehydes into beta-keto esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7772-99-8
189.61
AXZWODMDQAVCJE-UHFFFAOYSA-L
24479
[Cl-].[Cl-].[Sn++]
Cl2Sn
MFCD00011241
Stannous chloride
CHEBI:78067

Specifications

652°C
7772-99-8
250 g
Cl2Sn
Stannous chloride
AXZWODMDQAVCJE-UHFFFAOYSA-L
λ²-tin(2+) dichloride
24479
189.62
3.95 g/mL
247°C
Crystalline
99% min
MFCD00011241
Soluble in water,alkalies,alcohol,methy lethyl ketone,methyl acetate,and acetone.
[Cl-].[Cl-].[Sn++]
189.61
CHEBI:78067
Air sensitive; Hygroscopic
Tin(II) chloride, anhydrous
Videos
Safety and Handling

Safety and Handling

P234-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P390-P501c

H290-H302+H332-H314-H317-H335-H373

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 231-868-0

RTECSNumber : XP8700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only