missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin(II) trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03960306

 View more versions of this product

Catalog No. AA3960306


Explore available promotions

Description

Description

Tin(II) trifluoromethanesulfonate is a mild Lewis acid used for stereoselective aldol and Michael reactions; [2,3]-Wittig and Ireland-Claisen rearrangements, Horner-Wadsworth-Emmons reactions, asymmetric [3 + 2] cycloaddition, ring opening reactions and Friedel-Crafts reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tin(II) trifluoromethanesulfonate
(decomposition)
MFCD00191251
UN3288
tin ii trifluoromethanesulfonate
IACSYIHNPIQPMP-UHFFFAOYSA-M
molecular hydrogen;tin(2+);trifluoromethanesulfonate
131676109
97%
62086-04-8
CH4F3O3SSn+
5 g
Moisture sensitive
Insoluble in water.
[HH].[HH].C(F)(F)(F)S(=O)(=O)[O-].[Sn+2]
271.805
416.82
Powder

Chemical Identifiers

62086-04-8
271.805
IACSYIHNPIQPMP-UHFFFAOYSA-M
131676109
[HH].[HH].C(F)(F)(F)S(=O)(=O)[O-].[Sn+2]
CH4F3O3SSn+
MFCD00191251
tin ii trifluoromethanesulfonate
molecular hydrogen;tin(2+);trifluoromethanesulfonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H500

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only