missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin(II) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

$125.33 - $125.33

Chemical Identifiers

CAS 62086-04-8
Molecular Formula C2F6O6S2Sn
Molecular Weight (g/mol) 416.85
MDL Number MFCD00191251
InChI Key IACSYIHNPIQPMP-UHFFFAOYSA-M
Synonym tin ii trifluoromethanesulfonate
PubChem CID 131676109
IUPAC Name molecular hydrogen;tin(2+);trifluoromethanesulfonate
SMILES [HH].[HH].C(F)(F)(F)S(=O)(=O)[O-].[Sn+2]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC312530010
SDS View Documents
Thermo Scientific Chemicals
312530010
1 g Plastic bottle
Each for $125.33
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

62086-04-8
416.85
IACSYIHNPIQPMP-UHFFFAOYSA-M
131676109
[HH].[HH].C(F)(F)(F)S(=O)(=O)[O-].[Sn+2]
C2F6O6S2Sn
MFCD00191251
tin ii trifluoromethanesulfonate
molecular hydrogen;tin(2+);trifluoromethanesulfonate

Specifications

>300.0°C
62086-04-8
1 g
C2F6O6S2Sn
tin ii trifluoromethanesulfonate
IACSYIHNPIQPMP-UHFFFAOYSA-M
molecular hydrogen;tin(2+);trifluoromethanesulfonate
131676109
98%
Plastic bottle
White to Yellow
Powder
97.5% min. (ex Sn)
MFCD00191251
Solubility in water: soluble
[HH].[HH].C(F)(F)(F)S(=O)(=O)[O-].[Sn+2]
416.85
416.85
Authentic
Tin(II) trifluoromethanesulfonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only