missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin(IV) 2-ethylhexanoate diisopropoxide, 98% (metals basis), Thermo Scientific Chemicals

$59.89 - $211.84

Chemical Identifiers

CAS 288374-32-3
MDL Number MFCD00210604
Synonym tin iv 2-ethylhexanoate diisopropoxide, tin iv 2-ethylhexanoatediisopropoxide metalsbasis
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3916903
View Documents
Thermo Scientific Chemicals
03916903
1 g
Each for $59.89
 
AA3916906
View Documents
Thermo Scientific Chemicals
03916906
5 g
Each for $211.84
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

288374-32-3
MFCD00210604
tin iv 2-ethylhexanoate diisopropoxide, tin iv 2-ethylhexanoatediisopropoxide metalsbasis

Specifications

288374-32-3
1 g
tin iv 2-ethylhexanoate diisopropoxide, tin iv 2-ethylhexanoatediisopropoxide metalsbasis
98%
Tin(IV) 2-ethylhexanoate diisopropoxide
MFCD00210604
Moisture sensitive
523.08
Liquid
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361-H315-H319
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.

P201-P202-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P501c

H315-H319-H361d

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only