missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tin(IV) chloride, 1M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 428341000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
Tin(IV) chloride | |
100 mL | |
MFCD00011242 | |
15, 8901 | |
Solubility in water: reacts | |
Cl[Sn](Cl)(Cl)Cl | |
260.52 | |
260.52 | |
AcroSeal™ Glass Bottle | |
0.874 |
7646-78-8,142-82-5 | |
Cl4Sn | |
1°C | |
tin tetrachloride, tin iv chloride, stannane, tetrachloro, tin chloride, stannic chloride, tetrachlorotin, tin perchloride, tintetrachloride, tin iv tetrachloride, sncl4 | |
HPGGPRDJHPYFRM-UHFFFAOYSA-J | |
tetrachlorostannane | |
24287 | |
1M, exact strength on the certificate of analysis | |
Colorless solution | |
0.8740g/mL |
RUO – Research Use Only