Learn More
Tin(IV) isopropoxide isopropanol adduct, 98% (metals basis), Thermo Scientific Chemicals
$128.51 - $558.22
Description
Tin(IV) isopropoxide isopropanol adduct is used has a important tin precursor for the synthesis of tin oxide nanoparticles. Used in Indium Tin Oxide films, electrochemical and conducting properties and thin films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
132951-93-0 | |
MFCD00145408 |
Specifications
132951-93-0 | |
131°C (1.6 mmHg) | |
1 g | |
Soluble in hydrocarbons and warm isopropanol. Hydrolyzes with water. Reacts with alcohols,ketones and esters. | |
98% | |
Tin(IV) isopropoxide isopropanol adduct |
∼130°C (decomposition) | |
MFCD00145408 | |
Moisture sensitive | |
414.15 | |
Wet Solid | |
(metals basis) |
Safety and Handling
GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H318-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only