Tingley™ Workbrutes™ Steel-Toe Overshoes
The industry's first injection-molded PVC steel toe overshoe
$54.31 - $64.52
Specifications
Certifications/Compliance ASTM F2413-05 M/I/75/C/75.
Color Black, Yellow
For Use With (Application) Shipping, warehousing, baggage handling, food processing, mining, construction, general industry, transportation, onsite visitors, and temporary employees
Material PVC
Sole Features Cleated outsole, wedge sole
View More Specs
Products 3
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-999-343E
View Documents
Tingley Rubber Co
35211XL
X-Large
Pair for $54.31
19-999-343B
View Documents
Tingley Rubber Co
35211LG
Large
Pair for $54.31
19-999-343C
View Documents
Tingley Rubber Co
35211MD
Medium
Pair for $64.52
Description
Description
An affordable way to turn everyday shoes into safety footwear.
  • 100% waterproof, seamless construction protects entire shoe from damage caused by site chemicals, contaminants, and wear
  • Snug-fitting design and cleated outsole help prevent slips and falls
  • Special Frigiflex™ PVC compound provides stretch and longer wear; stays flexible in cold temperatures
  • Features include pull-on tab and heel kicker for easy on/off
Order Info
Also available in X-Small; contact your Fisher Safety Customer Service Representative for ordering information.
Compliance
Molded-in, oversized steel-toe cap accommodates most shoe styles and exceeds ASTM™ Safety Spec F 2413-05 M/I/75/C 75.
Specifications
Specifications
ASTM F2413-05 M/I/75/C/75.
Shipping, warehousing, baggage handling, food processing, mining, construction, general industry, transportation, onsite visitors, and temporary employees
Cleated outsole, wedge sole
Yes
Black, Yellow
PVC
Chemicals, fats, certain acids, hydrocarbons, caustics
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'