missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tiron, Thermo Scientific Chemicals

$78.59 - $219.79

Chemical Identifiers

CAS 149-45-1
Molecular Formula C6H10Na2O8S2
Molecular Weight (g/mol) 320.238
MDL Number MFCD00149531
InChI Key HEOKHLCODUWALT-UHFFFAOYSA-N
Synonym 4,5-Dihydroxy-1,3-benzenedisulfonic acid, disodium salt monohydrate
PubChem CID 131674010
IUPAC Name 4,5-dihydroxybenzene-1,3-disulfonic acid;molecular hydrogen;sodium
SMILES [HH].[HH].C1=C(C=C(C(=C1S(=O)(=O)O)O)O)S(=O)(=O)O.[Na].[Na]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC174140250
SDS View Documents
Thermo Scientific Chemicals
174140250
25 g Glass bottle
Each for $78.59
 
AC174141000
SDS View Documents
Thermo Scientific Chemicals
174141000
100 g Glass bottle
Each for $219.79
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

149-45-1
320.238
HEOKHLCODUWALT-UHFFFAOYSA-N
131674010
[HH].[HH].C1=C(C=C(C(=C1S(=O)(=O)O)O)O)S(=O)(=O)O.[Na].[Na]
C6H10Na2O8S2
MFCD00149531
4,5-Dihydroxy-1,3-benzenedisulfonic acid, disodium salt monohydrate
4,5-dihydroxybenzene-1,3-disulfonic acid;molecular hydrogen;sodium

Specifications

>300.0°C
C6H10Na2O8S2
MFCD00149531
15, 9621
Solubility in water: freely soluble.
Authentic
4,5-dihydroxybenzene-1,3-disulfonic acid;molecular hydrogen;sodium
131674010
332.21
White
Powder
149-45-1
(HO)2C6H2(SO3Na)2·H2O
11, 297
4,5-Dihydroxy-1,3-benzenedisulfonic acid, disodium salt monohydrate
HEOKHLCODUWALT-UHFFFAOYSA-N
[HH].[HH].C1=C(C=C(C(=C1S(=O)(=O)O)O)O)S(=O)(=O)O.[Na].[Na]
320.238
4% to 6% (typically)
Glass bottle
25 g
Tiron™
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only