missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
TISAB II, For Fluoride, Certified, LabChem™
Supplier: LabChem LC261304
Specifications
TISAB II | |
For Fluoride | |
7647-14-5,64-19-7,1310-73-2,125572-95-4 | |
Soluble in water | |
Certified | |
Poly Bottle | |
Carbon monoxide; Carbon dioxide | |
4 L |
Colorless | |
83.7,5.8,5.7,4.4,0.4 | |
CH3COOH | |
60.05 | |
Passes Test | |
Traceable to NIST | |
5.3 to 5.5 | |
Liquid |