Fisherbrand™ Tissue Path™ II Cassettes
Standard lids fit securely onto cassettes.
$417.38 - $670.13
Specifications
Closure Type Separate Lid
Material High Density Polymer
Product Type Tissue Cassette
View More Specs
Products 5
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
15-120-400D
View Documents
Fisherbrand™
58127
Blue
Case of 1500 Each for $417.38
15-120-400A
View Documents
Fisherbrand™
58128
Yellow
Case of 1500 Each for $670.13
15-120-400E
View Documents
Fisherbrand™
58126
White
Case of 1500 Each for $478.31
15-120-400C
View Documents
Fisherbrand™
58129
Green
Case of 1500 Each for $448.66
15-120-400B
View Documents
Fisherbrand™
SH58130
Pink
Case of 1500 Each for $502.94
Description
Description
  • Molded from a special high-density polymer resistant to all commonly used histological solvents
  • Slanted writing surface accepts markings easily, permitting positive sample identification
Order Info
Packaged in three dispenser boxes of 500
Specifications
Specifications
Separate Lid
High Density Polymer
Tissue Cassette
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'