Fisherbrand™ Tissue Path™ Microsette™ Six Compartment Biopsy Cassettes
Tissue Path™ Microsette™ Six Compartment Biopsy Cassettes are innovative, disposable and make biopsy pads obsolete.
Supplier:  Fisherbrand™ M5034
 View more versions of this product
Catalog No. 361041650
Explore available promotions
Description
Description
  • Molded from a special high-density polymer
  • Mesh design features more than 2000, 0.26 mm ''pores'' for maximum fluid exchange and drainage
  • One piece, integral lid eliminates the need for separate lids
  • Lids can be opened or closed as many times as needed and relocked securely each time
  • 45° anterior writing surface suitable for use with certain types of labeling instruments
  • Six-Compartment Cassette holds up to six tissue specimens; each one placed in a separate 0.25 x 0.43 in. (7 x 12 mm) numbered compartment
Specifications
Specifications
Mesh
Green
High Density Polymer
45°
6
Biopsy Cassette