missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(IV) chloride, 99.9%, Thermo Scientific Chemicals

$118.98 - $118.98

Chemical Identifiers

CAS 7550-45-0
MDL Number MFCD00011267
InChI Key XJDNKRIXUMDJCW-UHFFFAOYSA-J
Synonym titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4
PubChem CID 160960
IUPAC Name titanium(4+);tetrachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC197231000
SDS View Documents
Thermo Scientific Chemicals
197231000
100 mL Glass bottle
Each for $118.98
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Chemical Identifiers

7550-45-0
XJDNKRIXUMDJCW-UHFFFAOYSA-J
160960
[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
MFCD00011267
titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4
titanium(4+);tetrachloride

Specifications

136.5°C
7550-45-0
Liquid
99.85% min
TiCl4
01,1169; 02,414; 03,291; 04,507; 05,671; 06,590; 07,370; 08,483; 09,468; 10,401; 11,337; 12,94; 13,6; 14,111; 15,317; 16,332; 17,344
titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4
XJDNKRIXUMDJCW-UHFFFAOYSA-J
titanium(4+);tetrachloride
160960
99.9%
1.72
Titanium(IV) chloride
-24.0°C
Yellow
100 mL
Cl4Ti
MFCD00011267
15, 9634
Solubility in water: reacts forming hcl. Other solubilities: miscible with cold cond. hcl, soluble in alcohol (reacts, forming hcl), miscible with aliphatic hydrocarbons and, chlorinated aliphatic hydrocarbons
[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
189.71
189.71
Glass bottle
1.7200g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.
May cause respiratory irritation.
Reacts violently with water.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger

EINECSNumber : 231-441-9

RTECSNumber : XR1925000

TSCA : TSCA

Recommended Storage : note: product may darken on exposure to light

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only