missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals

$102.34 - $443.88

Chemical Identifiers

CAS 546-68-9
Molecular Formula C12H28O4Ti
Molecular Weight (g/mol) 284.26
InChI Key VXUYXOFXAQZZMF-UHFFFAOYSA-N
Synonym titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate
PubChem CID 11026
IUPAC Name propan-2-olate;titanium(4+)
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC194700050
SDS View Documents
Thermo Scientific Chemicals
194700050
5 mL Glass bottle
Each for $102.34
 
AC194702500
SDS View Documents
Thermo Scientific Chemicals
194702500
250 mL Glass bottle
Each for $102.63
 
AC194700010
SDS View Documents
Thermo Scientific Chemicals
194700010
1 L Glass bottle
Each for $228.05
 
AC194700025
SDS View Documents
Thermo Scientific Chemicals
194700025
2.5 L Glass bottle
Each for $443.88
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Chemical Identifiers

546-68-9
284.26
titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate
propan-2-olate;titanium(4+)
C12H28O4Ti
VXUYXOFXAQZZMF-UHFFFAOYSA-N
11026
CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4]

Specifications

546-68-9
0.9500g/mL
46°C
16.6 to 17.3% (Ti)
C12H28O4Ti
Ti[OCH(CH3)2]4
01,II,382
0.95
titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate
VXUYXOFXAQZZMF-UHFFFAOYSA-N
propan-2-olate;titanium(4+)
11026
284.26
Liquid
>15.0°C
240.0°C
Authentic
Glass bottle
1.4654 to 1.4684
5 mL
11,92; 12,90; 13,13; 14,61; 15,308; 16,54; 17,347
15,9636
Solubility in water: hydrolysis. Other solubilities: soluble in most common organic solvents
CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4]
284.26
4.3 mPa.s (25°C)
98+%
Titanium(IV) isopropoxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if inhaled.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remov

GHS Signal Word: Danger

EINECSNumber : 208-909-6

Recommended Storage : Product may darken during storage

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only