missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Titanium(IV) Oxide, Reag. Ph Eur, For Analysis, EMSURE™, MilliporeSigma™
Supplier: Technidata 1008081000
Description
For analysisSpecifications
Titanium(IV) oxide | |
Powder | |
O2Ti | |
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance | |
O=[Ti]=O | |
79.87 | |
CHEBI:32234 | |
Reagent Ph. Eur. | |
Plastic bottle |
13463-67-7 | |
1 kg | |
MFCD00011269 MFCD00210650 | |
GWEVSGVZZGPLCZ-UHFFFAOYSA-N | |
dioxotitanium | |
26042 | |
79.87 | |
Odorless | |
White powder |
Chemical Identifiers
13463-67-7 | |
79.87 | |
GWEVSGVZZGPLCZ-UHFFFAOYSA-N | |
26042 | |
dioxotitanium |
O2Ti | |
MFCD00011269 MFCD00210650 | |
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance | |
CHEBI:32234 | |
O=[Ti]=O |