Learn More
Titanium(IV) sulfide, 99.8% (metals basis), Thermo Scientific Chemicals
$595.33 - $2741.10
Chemical Identifiers
CAS | 12039-13-3 |
---|---|
Molecular Formula | S2Ti |
Molecular Weight (g/mol) | 111.99 |
MDL Number | MFCD00049598 |
InChI Key | CFJRPNFOLVDFMJ-UHFFFAOYSA-N |
Synonym | titanium sulfide tis2, titanium disulfide, titanium iv sulfide, bis sulfanylidene titanium, titanium disulphide, titansulfid, titanium sulfide, tis2, titanium iv sulfide-ti, titanium disulfide, crystal |
PubChem CID | 61544 |
SMILES | S=[Ti]=S |
Description
Titanium(IV) sulfide is used in solid lubricant, cathode material in rechargeable batteries, corrosion resistance alloys, solid-state lithium batteries, hybrid electric vehicles and plug-in electric vehicles. It is used in the synthesis of nanoclusters, nanodisks and nanotubes. It has high electrical conductivity. Titanium electrodes are used in chloralkali electrolysis, electrodialysis, galvanic technology, light bulbs, and bone pins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12039-13-3 | |
111.99 | |
CFJRPNFOLVDFMJ-UHFFFAOYSA-N | |
61544 |
S2Ti | |
MFCD00049598 | |
titanium sulfide tis2, titanium disulfide, titanium iv sulfide, bis sulfanylidene titanium, titanium disulphide, titansulfid, titanium sulfide, tis2, titanium iv sulfide-ti, titanium disulfide, crystal | |
S=[Ti]=S |
Specifications
12039-13-3 | |
5 g | |
S2Ti | |
titanium sulfide tis2, titanium disulfide, titanium iv sulfide, bis sulfanylidene titanium, titanium disulphide, titansulfid, titanium sulfide, tis2, titanium iv sulfide-ti, titanium disulfide, crystal | |
CFJRPNFOLVDFMJ-UHFFFAOYSA-N | |
disulfanylidenetitanium | |
61544 | |
Moisture sensitive |
Powder | |
99.8%, metal basis | |
MFCD00049598 | |
Titanium(IV) sulfide is insoluble in water and moderately acid soluble. | |
S=[Ti]=S | |
111.99 | |
112.03 | |
Titanium(IV) Sulfide |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
P235+P410-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H252-H315-H319-H335
DOTInformation : DOT Class: 4.2; Packaging Group: III
EINECSNumber : 234-883-0
RTECSNumber : XR2460000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only