missing translation for 'onlineSavingsMsg'
Learn More

Titanyl Phthalocyanine (purified by sublimation), TCI America™

$128.62 - $452.97

Chemical Identifiers

CAS 26201-32-1
Molecular Formula C32H18N8OTi
Molecular Weight (g/mol) 578.418
MDL Number MFCD00145414
InChI Key DOEAHNYJXVICHS-UHFFFAOYSA-N
Synonym TiOPc, Oxo(phthalocyaninato)titanium
PubChem CID 131664299
SMILES C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C([N-]4)N=C6C7=CC=CC=C7C(=N6)N=C8C9=CC=CC=C9C(=N8)N=C2[N-]3.O.[Ti+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T2272200MG
View Documents
TCI America
T2272200MG
200 mg
Each for $128.62
Only null left
 
T22721G
View Documents
TCI America
T22721G
1 g
Each for $452.97
Only null left
 
Description

Description

Specifications

Chemical Identifiers

26201-32-1
578.418
DOEAHNYJXVICHS-UHFFFAOYSA-N
131664299
C32H18N8OTi
MFCD00145414
TiOPc, Oxo(phthalocyaninato)titanium
C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C([N-]4)N=C6C7=CC=CC=C7C(=N6)N=C8C9=CC=CC=C9C(=N8)N=C2[N-]3.O.[Ti+2]

Specifications

26201-32-1
Blue-Purple
MFCD00145414
TiOPc, Oxo(phthalocyaninato)titanium
C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C([N-]4)N=C6C7=CC=CC=C7C(=N6)N=C8C9=CC=CC=C9C(=N8)N=C2[N-]3.O.[Ti+2]
131664299
Crystalline Powder
395°C
C32H18N8OTi
200 mg
DOEAHNYJXVICHS-UHFFFAOYSA-N
578.418
576.40
Titanyl Phthalocyanine (purified by sublimation)
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'