Titration Solvent, MilliporeSigma™
Solvent for volumetric Karl Fischer titration with two component reagents Aquastar™.
Supplier:  Technidata 1880152500
 View more versions of this product
Catalog No. M1880152500
Explore available promotions
Description
Description
Specifications
Specifications
11°C (Methanol)
2.5 L
0.90 g/cm3 (at 20°C)
Glass Bottle