Supelco TLC Explorer
The TLC Explorer Documentation System is designed to digitally evaluate and record thin-layer chromatography (TLC), boosting the efficiency and accuracy of chromatography workflows.
Supplier:  Supelco 1526100001
Catalog No. 15-261-00001
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • The TLC Explorer Documentation System is designed to digitally evaluate and record thin-layer chromatography (TLC), boosting the efficiency and accuracy of chromatography workflows.
  • It provides precise TLC imaging for reliable densitometric measurement, facilitating quantitative analysis and consistent data interpretation.
  • Moreover, the intuitive software integrates seamlessly with existing lab systems, enabling efficient data sharing and documentation.
Specifications
Specifications
LED
Thin Layer Chromatography
TLC Documentation System