MilliporeSigma™ TLC Silica Gel 60, F254, MS-Grade Glass Plates, 20x20
MS-Grade Glass TLC plates (20 x 20cm) with Silica gel 60 coated with fluorescent indicator F254, 25/Box
Supplier: MilliporeSigma™ 1009330001
Specifications
60 Å | |
F254 | |
20 cm |
Glass TLC Plate | |
Silica Gel | |
20 cm |