missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals TMB QUICK, for ELISA, Liquid Substrate

$244.85 - $348.66

Specifications

Boiling Point 100°C
Color Clear
Form Liquid
Quantity 100mL
Substrate Type TMB Substrate
Packaging Glass Bottle
Substrate Quantity 100mL

Products 2
Catalog Number Mfr. No. Substrate Quantity Price Quantity  
Catalog Number Mfr. No. Substrate Quantity Price Quantity  
AC328051000
SDS View Documents
Thermo Scientific Chemicals
328051000
100mL
Each for $348.66
 
AC328050500
SDS View Documents
Thermo Scientific Chemicals
328050500
50mL
Each for $244.85
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

100°C
Liquid
TMB Substrate
100mL
Clear
100mL
Glass Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only