missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ TMSI Silylation Reagent

Employ the strongest silylator available for carbohydrates and steroids with the Thermo Scientific™ TMSI Silylation Reagent.

$577.88 - $1226.32

Specifications

Standard Concentration 98 to 100%
For Use With GC/MS
Hazards Flammable, Irritant
Storage 2°C to 8°C
Standard Components 1-(Trimethylsilyl)-1 H-imidazole
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PI88623
View Documents
Thermo Scientific™
TS88623
10 x 1 mL ampules
Pack of 10 Each for $577.88
 
PI88625
View Documents
Thermo Scientific™
TS88625
25 g
Pack for $1,226.32
 
Description

Description

Employ the strongest silylator available for carbohydrates and steroids with the Thermo Scientific™ TMSI Silylation Reagent. This silylation reagent reacts quickly with hydroxyls and carboxylic acids but not with amines, and is particularly well suited in multiderivatization schemes for compounds containing both hydroxyl and amine groups. TMSI may be used to derivatize alcohols, phenols, organic acids, hormones, glycols, nucleotides, and narcotics and is excellent for C1–C5 fatty acids in serum and urine.

  • Reacts quickly and smoothly with hydroxyls and carboxylic acids but not with amines
  • Especially useful in multiderivatization schemes for compounds containing both hydroxyl and amine groups
  • Used in the derivatization of alcohols, phenols, organic acids, steroids, hormones, glycols, nucleotides and narcotics
  • Excellent for C1 through C5 fatty acids in serum and urine
Specifications

Specifications

98 to 100%
Flammable, Irritant
1-(Trimethylsilyl)-1 H-imidazole
GC/MS
2°C to 8°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.