missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tobramycin, 900μg/mg, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J6604006
Description
Primarily targets Gram-negative bacteria
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tobramycin | |
Plastic bottle | |
MFCD00077885 | |
1357507 | |
Soluble in water | |
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(CC(C(O3)CN)O)N)N | |
467.52 | |
CHEBI:28864 | |
Powder |
32986-56-4 | |
C18H37N5O9 | |
5 g | |
tobramycin, nebramycin 6, nebramycin factor 6, nebramycin vi, tobramicin, tobramycetin, nebcin, 3'-deoxykanamycin b, aktob, tobi | |
NLVFBUXFDBBNBW-PBSUHMDJSA-N | |
(2S,3R,4S,5S,6R)-4-amino-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-6-(hydroxymethyl)oxane-3,5-diol | |
36294 | |
467.51 |
Chemical Identifiers
32986-56-4 | |
467.52 | |
NLVFBUXFDBBNBW-PBSUHMDJSA-N | |
36294 | |
(2S,3R,4S,5S,6R)-4-amino-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-6-(hydroxymethyl)oxane-3,5-diol |
C18H37N5O9 | |
MFCD00077885 | |
tobramycin, nebramycin 6, nebramycin factor 6, nebramycin vi, tobramicin, tobramycetin, nebcin, 3'-deoxykanamycin b, aktob, tobi | |
CHEBI:28864 | |
C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(CC(C(O3)CN)O)N)N |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 251-322-5
RTECSNumber : WK2100000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only