TOC Standard, According to ISO/CEN EN1484, 25 mg/L±2 mg/L, MilliporeSigma™ Supelco™
TOC standard can be used in the analysis of degradation by-products of acetaminophen (ACE) solution during electro-Fenton (EF) oxidation performed on carbon felt.
Supplier: Merck Emd Millipore 90326250MLF
Specifications
25 mg/L ±2 mg/L | |
250 mL | |
Analytical Standard |