missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Toluene, 99.5%, for spectroscopy ACS, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 421170025

 View more versions of this product

Catalog No. AC421170025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
Specifications

Specifications

Toluene
99.5%
0.01 max. at 350nm, 0.01 max. at 400nm, 0.02 max. at 335nm, 0.05 max. at 310nm, 0.1 max. at 300nm, 0.2 max. at 293nm, 0.5 max. at 288nm, 1 max. at 286nm
111.0°C
4°C
Liquid
0.001% max.
0.866
C6H5CH3
Vapor Pressure: 29mbar at 20°C
0.03% max. (Coulometric)
15, 9688
Solubility in water: almost insoluble (0.5g/L at 20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, acetone,, ether, petroleum ether, benzene, ch2cl2, dmf, gla, cial acetic acid, chcl3, ccl4.
CC1=CC=CC=C1
92.14
CHEBI:17578
99.5%
-95.0°C
108-88-3
0.8660g/mL
2.5 L
Glass Bottle
1.4968
99.5%
C7H8
MFCD00008512
0.6 mPa.s (20°C)
05, 280
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
YXFVVABEGXRONW-UHFFFAOYSA-N
toluene
1140
92.14
ACS

Chemical Identifiers

108-88-3
92.14
YXFVVABEGXRONW-UHFFFAOYSA-N
1140
toluene
C7H8
MFCD00008512
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
CHEBI:17578
CC1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. May be fatal if swallowed and enters airways. Causes skin irritation. Suspected of damaging the unborn child. Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.

Danger

EINECSNumber : 203-625-9

RTECSNumber : XS5250000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only