Learn More
Toluene, 99.85%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
$80.98 - $244.51
Chemical Identifiers
CAS | 108-88-3 |
---|---|
Molecular Formula | C7H8 |
Molecular Weight (g/mol) | 92.14 |
MDL Number | MFCD00008512 |
InChI Key | YXFVVABEGXRONW-UHFFFAOYSA-N |
Synonym | methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene |
PubChem CID | 1140 |
ChEBI | CHEBI:17578 |
IUPAC Name | toluene |
SMILES | CC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AC364411000
|
Thermo Scientific Chemicals
364411000 |
100 mL |
Each for $80.98
|
|
||||||
AC364415000
|
Thermo Scientific Chemicals
364415000 |
500 mL |
Each for $99.92
|
|
||||||
AC364410010
|
Thermo Scientific Chemicals
364410010 |
1 L |
Each for $120.94
|
|
||||||
AC364410025
|
Thermo Scientific Chemicals
364410025 |
2.5 L |
Each for $244.51
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-88-3 | |
92.14 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
1140 | |
toluene |
C7H8 | |
MFCD00008512 | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
CHEBI:17578 | |
CC1=CC=CC=C1 |
Specifications
-95.0°C | |
Colorless | |
111.0°C | |
4°C | |
Liquid | |
99.85% | |
C7H8 | |
MFCD00008512 | |
0.6 mPa.s (20°C) | |
05, 280 | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
toluene | |
1140 | |
92.14 | |
Extra Dry over Molecular Sieve |
108-88-3 | |
0.8660g/mL | |
100 mL | |
Authentic | |
1.4968 | |
0.866 | |
C6H5CH3 | |
Vapor Pressure: 29mbar at 20°C | |
0.005% Max. (Coulometric) | |
15, 9688 | |
Solubility in water: almost insoluble (0.5g/L at 20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, acetone,, ether, petroleum ether, benzene, ch2cl2, dmf, gla, cial acetic acid, chcl3, ccl4. | |
CC1=CC=CC=C1 | |
92.14 | |
CHEBI:17578 | |
99.85% | |
Toluene, water <50 ppm |
Safety and Handling
GHS P Statement May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. May be fatal if swallowed and enters airways. Causes skin irritation. Suspected of damag
Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0
Signal Word: DANGER!
RUO – Research Use Only