missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Toluene, ACS Grade, ≥ 99.5 %, LabChem™
$161.00 - $551.21
Chemical Identifiers
CAS | 108-88-3 |
---|---|
Molecular Formula | C7H8 |
Molecular Weight (g/mol) | 92.14 |
MDL Number | MFCD00008512 |
InChI Key | YXFVVABEGXRONW-UHFFFAOYSA-N |
Synonym | methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene |
PubChem CID | 1140 |
ChEBI | CHEBI:17578 |
IUPAC Name | toluene |
SMILES | CC1=CC=CC=C1 |
Chemical Identifiers
108-88-3 | |
92.14 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
1140 | |
toluene |
C7H8 | |
MFCD00008512 | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
CHEBI:17578 | |
CC1=CC=CC=C1 |
Specifications
-95°C | |
Colorless | |
Carbon dioxide; Carbon monoxide | |
111°C | |
Passes Test | |
Liquid | |
870kg/m3 | |
C7H8 | |
0.69mm2/s | |
≤0.03% | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
toluene | |
1140 | |
92.14 | |
ACS |
108-88-3 | |
100 | |
870kg/m3 | |
4 L | |
Amber Glass | |
≤0.001% | |
C7H8 | |
MFCD00008512 | |
UN1294 | |
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene | |
CC1=CC=CC=C1 | |
92.14 | |
CHEBI:17578 | |
≥99.5% | |
Toluene |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Suspected of damaging fertility or the unborn child.
May cause damage
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/at
Danger
EINECSNumber : 203-625-9
Recommended Storage : Room Temperature